Learn More
BA Antigen (Leu21) Antibody (DF1588) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | BA Antigen (Leu21) |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Clone | DF1588 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, CyTOF-ready |
| Formulation | PBS with No Preservative |
| Host Species | Mouse |
| Immunogen | Human leucocytes |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.