Learn More
Baculovirus Envelope gp64 protein Antibody (AcV1), DyLight 680, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Baculovirus Envelope gp64 protein |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AcV1 |
| Conjugate | DyLight 680 |
| Dilution | Flow Cytometry |
| Gene Alias | ACNVgp129, AcOrf-128 |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody is baculovirus envelope gp64 protein. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.