Learn More
Stoelting™ Barnes Maze
SDP
Barnes Maze task is similar to the Radial Arm and Morris Water mazes but without dietary restrictions and with less physical stress.
$94.50 - $5950.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 6
Catalog Number Mfr. No. Animal Includes Price Quantity & Availability  
Catalog Number Mfr. No. Animal Includes Price Quantity & Availability  
10-000-305
SDP
View Documents
Stoelting™
60170
Mouse
Each for $5,330.00
Only null left
Add to Cart
10-000-607
SDP
View Documents
Stoelting™
60170F
Mouse False Box
Each for $94.50
Only null left
Add to Cart
10-000-608
SDP
View Documents
Stoelting™
60170G
Mouse Goal Box
Each for $197.70
Only null left
Add to Cart
10-000-328
SDP
View Documents
Stoelting™
60270
Rat
Each for $5,950.00
Only null left
Add to Cart
10-000-609
SDP
View Documents
Stoelting™
60270F
Rat False Box
Each for $136.30
Only null left
Add to Cart
10-000-610
SDP
View Documents
Stoelting™
60270G
Rat Goal Box
Each for $197.70
Only null left
Add to Cart
Description
Description
  • Designed for testing spatial learning and memory.
  • Within the maze, animals are motivated to escape from the brightly lit, open platform to a small dark recessed chamber (escape box).
  • During testing, the animal learns the spatial location of the escape box.
  • Optional false escape boxes help to remove visual cues that might be observed from a distance or through an open hole.
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.