Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2L10 Antibody (8A11G12) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200080755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61704 Supplier Novus Biologicals Supplier No. NBP261704
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCL2L10 Monoclonal specifically detects BCL2L10 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen BCL2L10
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 8A11G12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Apoptosis regulator Bcl-B, apoptotic protein NrH, bcl2-L-10, BCL2-like 10 (apoptosis facilitator), bcl-2-like protein 10, BCLB, BCL-B, Boo, Diva, MGC129810, MGC129811
Gene Symbols BCL2L10
Host Species Mouse
Immunogen Purified recombinant fragment of human BCL2L10 (AA: 31-186) expressed in E. Coli.
Molecular Weight of Antigen 22 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10017
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.