Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Far-red fluorescent DNA dye
- Cell permeable
- Used to stain nucleic acids in live or fixed cells
- Has an excitation wavelength maxima of 598/646nm
Specifications
Specifications
| Color | Far-red |
| Concentration | 5 mM |
| Content And Storage | Store undiluted at 4°C |
| For Use With (Application) | Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry) |
| Dye Type | DNA dye |
| Excitation Wavelength Range | 598/646 nm |
| Quantity | 50 μL |
| Reactivity | Tested in Development: Human, Mouse; Reported Reactivity: Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction