Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclin 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$206.50 - $499.50
Specifications
Antigen | Beclin 2 |
---|---|
Concentration | 0.9 mg/mL |
Dilution | Western Blot 1:200-1:500, Simple Western 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26506105) |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11060984T
![]() |
Novus Biologicals
NB11060984SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11060984
![]() |
Novus Biologicals
NB11060984 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Beclin 2 Polyclonal specifically detects Beclin 2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Beclin 2 | |
Western Blot 1:200-1:500, Simple Western 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin reported in scientific literature (PMID 26506105) | |
Polyclonal | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Tumor Suppressors | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 | |
BECN2 | |
IgG | |
Affinity Purified | |
50 kDa |
0.9 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
A8MW95 | |
441925 | |
A synthetic peptide made to an internal region (within residues 200-300) of the human Beclin 2 protein. [UniProt# A8MW95] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title