Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenesulfonic acid isopropyl ester, 95%

Catalog No. AAH6452306
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 6214-18-2 | C9H12O3S | 200.252 g/mol

A sulfonate ester that acts as a potential effects in drug substances impurity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6214-18-2
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.252
MDL Number MFCD04038771
InChI Key YQZZXXKFKTWDPY-UHFFFAOYSA-N
PubChem CID 12441430
IUPAC Name propan-2-yl benzenesulfonate
SMILES CC(C)OS(=O)(=O)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Refractive Index 1.502
Quantity 5 g
Beilstein 1106314
Synonym isopropyl benzenesulfonate, isopropylbenzenesulfonate, benzenesulfonic acid, 1-methylethyl ester, benzenesulfonic acid isopropyl ester, isopropyl besylate, benzenesulfonic acid,1-methylethyl ester, benzenesulfonic acid 1-methylethyl ester
Solubility Information Soluble in chloroform and ethyl acetate.
Formula Weight 200.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Benzenesulfonic acid isopropyl ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DB7151080
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.