Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylamine, 99%, pure

Catalog No. p-65971
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

Catalog No. AC105851000

Supplier: Thermo Scientific Chemicals 105851000

Only null left
Add to Cart

CAS: 100-46-9 | C7H9N | 107.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.15
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Colorless to Light Yellow
Density 0.9800g/mL
Boiling Point 182.0°C to 185.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5424 to 1.5444
Linear Formula C6H5CH2NH2
Quantity 100 mL
Beilstein 12,1013
Fieser 01,51; 10,26; 13,79; 16,22
Merck Index 15,1128
Specific Gravity 0.98
Solubility Information Solubility in water: soluble in water. Other solubilities: >50g/100mL methanol,>50g/100mL ethanol,>50g/100mL 2-propanol,miscible with alcohol,ether,acetone,benzene
Viscosity 1.82 mPa.s (20°C)
Formula Weight 107.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-854-1
RTECSNumber DP1488500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.