Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltributylammonium bromide, 99%, Thermo Scientific™

Catalog No. AC222890050
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
50 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-59-0
MDL Number MFCD00011848
InChI Key UDYGXWPMSJPFDG-UHFFFAOYSA-M
Synonym benzyltributylammonium bromide, tributylbenzylammonium bromide, n-benzyl-n,n-dibutylbutan-1-aminium bromide, benzyl tributyl ammonium bromide, benzenemethanaminium, n,n,n-tributyl-, bromide, benzyltri-n-butylammonium bromide, benzyltributylazanium bromide, benzyl-tributyl-azanium, benzyltributylammoniumbromide, pubchem7801
PubChem CID 2724282
IUPAC Name benzyl(tributyl)azanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1.[Br-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 168.0°C to 174.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C19H34BrN
Linear Formula C6H5CH2N[(CH2)3CH3]3Br
Quantity 5 g
Molecular Weight (g/mol) 356.38
Formula Weight 356.38
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Benzyltributylammonium bromide, 99%
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 246-819-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.