Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bestrophin 1 Antibody (E6-6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 42 publications
$258.08 - $615.90
Specifications
| Antigen | Bestrophin 1 |
|---|---|
| Clone | E6-6 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 30048622), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24345323), Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 27519691), Knockout Validated, Dual RNAscope ISH-IHC |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300164SS
![]() |
Novus Biologicals
NB300164SS |
0.025 mL |
Each for $258.08
|
|
|||||
NB300164
![]() |
Novus Biologicals
NB300164 |
0.1 mL |
Each for $615.90
|
|
|||||
Description
Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Human, Mouse, Porcine, Canine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dual RNAscope ISH-IHC.Specifications
| Bestrophin 1 | |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Sensory Systems, Vision | |
| PBS with 0.02% Sodium Azide | |
| ARB, BEST, Best disease, bestrophin 1, bestrophin-1, BMD, TU15B, vitelliform macular dystrophy 2, Vitelliform macular dystrophy protein 2, VMD2RP50 | |
| BEST1 | |
| IgG1 κ | |
| Protein A or G purified | |
| Bestrophin (E6-6) |
| E6-6 | |
| Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 30048622), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24345323), Immunohistochemistry-Frozen, Proximity Ligation Assay reported in scientific literature (PMID 27519691), Knockout Validated, Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Pig, Canine, Primate, Rat (Negative) | |
| O76090 | |
| 7439 | |
| Synthetic peptide conjugated to KLH corresponding to the C-terminus of human Bestrophin 1 (KDHMDPYWALENRDEAHS) [Uniprot: O76090] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title