Learn More
Description
Specifications
Specifications
| Antigen | beta Tubulin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:1000. Use reported in scientific literature (PMID 35008713), Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:1000 - 1:2000 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | P07437 |
| Gene Alias | beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta |
| Gene Symbols | TUBB |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
