Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-1,4-Galactosyltransferase 2/B4GalT2 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108357 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159864 100 μL
NBP15986420 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP159864 Supplier Novus Biologicals Supplier No. NBP159864
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

beta-1,4-Galactosyltransferase 2/B4GalT2 Polyclonal specifically detects beta-1,4-Galactosyltransferase 2/B4GalT2 in Human samples. It is validated for Western Blot.

Specifications

Antigen beta-1,4-Galactosyltransferase 2/B4GalT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O60909
Gene Alias B4Gal-T2, B4Gal-T3, beta-1,4-galactosyltransferase 2, Beta-1,4-GalTase 2, beta-4-GalT2, beta4Gal-T2, beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 2, EC 2.4.1.-, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 2, UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2, UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 2, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 2, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 2
Gene Symbols B4GALT2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to B4GALT2(UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 2) The peptide sequence was selected from the middle region of B4GALT2. Peptide sequence NRISLTGMKISRPDIRIGRYRMIKHDRDKHNEPNPQRFTKIQNTKLTMKR The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 42 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8704
Test Specificity Goat 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.