Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-1 Adrenergic R/ADRB1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600978 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600978 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600978 Supplier Novus Biologicals Supplier No. NB600978
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

beta-1 Adrenergic R/ADRB1 Polyclonal specifically detects beta-1 Adrenergic R/ADRB1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay, Peptide ELISA.

Specifications

Antigen beta-1 Adrenergic R/ADRB1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In vitro Assay, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 4 - 6 ug/mL, In vitro assay, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P08588
Gene Alias ADRB1RRHR, adrenergic, beta-1-, receptor, B1AR, beta-1 adrenergic receptor, Beta-1 adrenoceptor, Beta-1 adrenoreceptor, BETA1AR
Gene Symbols ADRB1
Host Species Goat
Immunogen Peptide with sequence ESDEARRCYNDPK corresponding to internal region according to NP_000675.1.
Molecular Weight of Antigen 51 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 153
Test Specificity Predicted cross-reactivity based on sequence identity: Dog, Mouse.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.