Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ beta Synuclein Monoclonal Antibody (6A10)
Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | beta Synuclein |
---|---|
Clone | 6A10 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547356
|
Invitrogen™
MA547356 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547455
|
Invitrogen™
MA547455 |
100 μL |
Each for $526.00
|
|
|||||
Description
beta Synuclein Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.Specifications
beta Synuclein | |
1 mg/mL | |
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
P33567, Q16143, Q63754, Q91ZZ3 | |
100127169, 104069, 113893, 508421, 6620 | |
C-terminal peptide of human beta-synuclein EPEGESYEDPPQEEYQEYEPEA coupled to KLH. | |
Antibody |
6A10 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Mouse, Rat, Bovine, Pig | |
SNCB | |
AI838531; betaSYN; beta-synuclein; Phosphoneuroprotein 14; PNP 14; Sncb; synuclein beta; synuclein, beta | |
SNCB | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title