Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns
Click to view available options
Length (Metric):
30 mm
Description
Outstanding size separation of water soluble compounds.
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 60Å pore size
- Particle size: 5μm
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 7.8 mm |
| For Use With | Size Exclusion Chromatography |
| Length (Metric) | 30 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| Pore Size | 60 Å |
| USP Type | L33 |
| Stationary Phase | Diol (OH) |
| Product Line | BioBasic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction