Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ BioBasic™ 8 5μm Nanobore HPLC Columns
Click to view available options
Length (Metric):
50 mm
100 mm
Description
At flow rates of nL/min versus mL/min, Thermo Scientific™ BioBasic 8 5μm Nanobore HPLC Columns offer greater signal to noise ratio than traditional columns used with electrospray.
- High sensitivity flexible silica capillary columns
- Optimized for LC/MS applications
Features of BioBasic 8 Solid Phase
- 300Å pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Optimized for LC/MS separations
Ordering Information:
Unless otherwise specified, IntegraFrit and PicoFrit are sold in single-column units.

Specifications
Specifications
Column Format | Nanobore Column with PicoFrit |
Diameter (Metric) | 75 μm |
For Use With | Nanospray LC/MS |
Length (Metric) | 100 mm |
Particle Size | 5 μm |
pH | 2 to 10 |
Pore Size | 300 Å |
USP Type | L7 |
Stationary Phase | C8 |
Packing Material | Silica |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction