Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma BIOshell A160 Peptide PCS-C18 HPLC Column
Shop All MilliporeSigma Supelco Products
Click to view available options
Diameter (Metric):
1.5 mm
2.1 mm
3.0 mm
4.6 mm
Length (Metric):
5 mm
50 mm
100 mm
150 mm
Description
These columns contain advanced Fused-Core™ particles with pores strategically sized to 160 Å to enable optimized peptide diffusion. This attribute makes these columns an excellent choice for peptide mapping.
- The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic peptides as compared to traditional C18 chemistries.
- In addition, the PCS-C18 facilitates the use of mobile phases with lower concentrations of mobile phase modifiers without any compromise to column performance.
- This column also provides orthogonal selectivity to ES-C18, ES-CN (cyano), and phenyl-hexyl chemistries.

Specifications
Specifications
Diameter (Metric) | 2.1 mm |
Length (Metric) | 150 mm |
Particle Size | 2.7 μm |
Pore Size | 160 Å |
Phase | Positively Charged Surface (PCS) |
Material | Stainless Steel |
Type | HPLC Column |
Technique | HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction