Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (BTN/2032R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$610.00
Specifications
| Antigen | Biotin (Vitamin B7 or Vitamin H) |
|---|---|
| Clone | BTN/2032R |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In-situ Hybridization |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB261393100
![]() |
Novus Biologicals
NBP261393100UG |
100 μg |
Each for $610.00
|
|
|||||
NB261393200
![]() |
Novus Biologicals
NBP261393200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal specifically detects Biotin (Vitamin B7 or Vitamin H) in All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, In-situ Hybridization, CyTOF-ready.Specifications
| Biotin (Vitamin B7 or Vitamin H) | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| Unconjugated | |
| Mouse | |
| 10mM PBS - BSA free with No Preservative | |
| IgG1 | |
| Protein A or G purified | |
| It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise. |
| BTN/2032R | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In-situ Hybridization | |
| Monoclonal | |
| Purified | |
| All species | |
| Biotinylated sheep immunoglobulin | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 2.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title