Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyethyl) phthalate, 96%, Thermo Scientific Chemicals

Catalog No. p-7059156
Click to view available options
Quantity:
50 g

Catalog No. AA4408918

Supplier: Thermo Scientific Chemicals 04408918

Only null left
Add to Cart

It finds it application in cosmetic industry. It is also used as a plasticizer for toys, including inflatable water products, hoppers, play and exercise balls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-82-8
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.29
MDL Number MFCD00042842
InChI Key HSUIVCLOAAJSRE-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl phthalate, kesscoflex mcp, 2-methoxyethyl phthalate, dmep, bis methoxyethyl phthalate, di 2-methoxyethyl phthalate, kodaflex dmep, methyl glycol phthalate, dimethoxyethyl phthalate, dimethylglycol phthalate
PubChem CID 8344
IUPAC Name bis(2-methoxyethyl) benzene-1,2-dicarboxylate
SMILES COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC

Specifications

Color Colorless
Density 1.17 g/mL
Boiling Point 340°C
Flash Point 185°C (365°F)
Refractive Index 1.503
Quantity 50 g
Beilstein 2056929
Solubility Information Soluble in water 8500mg/L at 25°C.
Formula Weight 282.29
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bis (2-methoxyethyl) phthalate
Show More Show Less
Hazard Category H360Df
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 204-212-6
RTECSNumber TI1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.