Learn More
Eisco™ Bituminous Coal Specimen
SCI_ED
Catalog No. p-7076834 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24720 1 Ea. 2 to 3 cm
S26604 10/Pk. 2 to 3 cm
S24721 12/Pk. 2 to 3 cm
S26659 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24720 Supplier Eisco™ Supplier No. ESNG0030
Only null left
Add to Cart
Add to Cart
Bituminous Coal is formed in anoxic environments from compressed organic matter, and is used worldwide as a fuel source.
  • Bituminous coal is mined heavily around world
  • Students can get close look at material that powers world
  • Has vitreous sheen, distinguishing it from other coals and sedimentary rocks
  • For use in lessons about unique anoxic conditions under which bituminous coal was formed
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Sedimentary Rock, Bituminous Coal
For Use With (Application) Lessons about unique anoxic conditions under which bituminous coal was formed
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm