Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Black Hematite Mineral Specimen
SCI_ED

Catalog No. p-7164877
Click to view available options
Quantity:
1 Ea.
10/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Hematite specimen that is ideal for demonstrations in mineral physical properties.

  • Black in color
  • Takes the form of tiny, sparkling iron oxide crystals
  • Correlates well to Next Generation Science Standards Evidence Statements

Specifications

Product Type Metamorphic Rock, Hematite
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.