Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IB/ALK-6 Antibody (477914), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312152 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312152 Supplier Novus Biologicals Supplier No. FAB5051APCCY7
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMPR-IB/ALK-6 Monoclonal antibody specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen BMPR-IB/ALK-6
Applications Flow Cytometry
Classification Monoclonal
Clone 477914
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6, Lys14-Arg126, Accession # O00238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 658
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.