Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bone marrow stromal cell antigen 1/CD157 Antibody (534509), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB369090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369090 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369090 Supplier Novus Biologicals Supplier No. FAB47361Z
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF

Specifications

Antigen Bone marrow stromal cell antigen 1/CD157
Applications Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 534509
Conjugate DyLight 755
Dilution Flow Cytometry, ELISA, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CD157, Gly29-Lys292, Accession # Q10588
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 683
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.