Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Bottle Top Dispenser with Bottle

Shop All United Scientific Supplies, Inc Products


Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 30 mL
5 to 60 mL
Increments:
0.05 mL
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
Weight (English):
1.5 lb.
1.75 lb.
2 lb.
2.25 lb.
4 lb.
Volume (Metric) Reservoir:
100 mL
250 mL
500 mL
1000 mL
2000 mL
Description
United brand bottle top dispensers offer high accuracy at economical prices.
- Each dispenser is individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
- A set of adapters to fit most laboratory reagent bottles are provided
- All wetted parts are made from PTFE and borosilicate glass for maximum chemical resistance
- These easy-to-use instruments are suitable for all laboratory applications, except HF
- Special precautions must be taken for use, cleaning, and storage when working with concentrated fuming acids
- A factory calibration certificate is provided with each bottle top dispenser
- Dispensers are easy to disassemble and can be recalibrated in the lab
- Fully autoclavable at 121°C, 15 psi for 10 to 15 minutes
Specifications
Specifications
| Accuracy | 0.005 |
| Product Type | Analog Bottletop Dispenser with Adapters |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 8655 |
| Color | White |
| Channels | 1 |
| Material | PTFE, Borosilicate Glass |
| Volume (Metric) | 1 to 10 mL |
| Display | Analog |
| For Use With (Application) | Dispensing Liquids |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction