Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD8 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB247063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB247063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB247063 Supplier Novus Biologicals Supplier No. NB10079780C
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BRD8 Polyclonal antibody specifically detects BRD8 in Human samples. It is validated for Western Blot

Specifications

Antigen BRD8
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bromodomain containing 8, bromodomain-containing protein 8, Skeletal muscle abundant protein, SMAP2, SMAPp120Skeletal muscle abundant protein 2, Thyroid hormone receptor coactivating protein of 120 kDa, trCP120
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 25 and 75 of human Bromodomain containing 8 using the numbering given in TrEMBL entry Q969M9 (GeneID 10902).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 10902
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.