Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Breast cancer suppressor candidate 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Breast cancer suppressor candidate 1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Breast cancer suppressor candidate 1 Polyclonal specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Breast cancer suppressor candidate 1 | |
Polyclonal | |
Rabbit | |
Human | |
BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A | |
VWA5A | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
4013 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGM | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title