Learn More
BriskHeat™ Lower Hemisphere Mantles for Flasks, No Controller
Uniform heat distribution for round-bottom flasks
$419.50 - $1591.00
Catalog Number | Mfr. No. | Capacity | Voltage | Wattage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Voltage | Wattage | Price | Quantity & Availability | |||
10-067-026
![]() |
BriskHeat™
HM0050HS1 |
50mL (1.6 oz.) | 120 V | 60w |
Each for $553.00
|
|
|||
10-067-027
![]() |
BriskHeat™
HM0100HS1 |
100mL (3.3 oz.) | 120 V | 80w |
Each for $576.50
|
|
|||
10-067-028
![]() |
BriskHeat™
HM0125HS1 |
125mL (4.2 oz.) | 120 V | 80w |
Each for $581.00
|
|
|||
10-067-030
![]() |
BriskHeat™
HM0250HS1 |
250mL (8.4 oz.) | 120 V | 180w |
Each for $620.00
|
|
|||
10-067-029
![]() |
BriskHeat™
HM0200HS1 |
200mL (6.7 oz.) | 120 V | 100w |
Each for $571.50
|
|
|||
10-067-033
![]() |
BriskHeat™
HM2000HS1 |
2000mL (67.6 oz.) | 120 V | 500w |
Each for $843.00
|
|
|||
10-067-032
![]() |
BriskHeat™
HM1000HS1 |
1000mL (33.8 oz.) | 120 V | 380w |
Each for $724.00
|
|
|||
10-067-035
![]() |
BriskHeat™
HM5000HS1 |
5000mL (169 oz.) | 120 V | 600w |
Each for $965.00
|
|
|||
10-067-031
![]() |
BriskHeat™
HM0500HS1 |
500mL (17 oz.) | 120 V | 270w |
Each for $684.00
|
|
|||
10-067-034
![]() |
BriskHeat™
HM3000HS1 |
3000mL (101.4 oz.) | 120 V | 500w |
Each for $909.50
|
|
|||
10-067-036
![]() |
BriskHeat™
HM12000HS1 |
12000mL (405.7 oz.) | 120 V | 1300w |
Each for $1,591.00
|
|
|||
10-067-038
![]() |
BriskHeat™
HM0050HS2 |
50mL (1.6 oz.) | 240 V | 60w |
Each for $419.50
|
|
|||
Description
Uniform heat distribution for round-bottom flasks
Lower hemispherical heating mantles are:
Ideal for use with basket ring stand
- Fiberglass cloth outer shell
- Uniform, low-watt density knitted and braided heating element. Temperatures up to 842°F (450°C).
- Multi-stranded resistance wire for increased durability and flexibility
- Temperature controller required
120VAC models include:
3 in. (76mm) lead wire with NEMA L1-15R connector and a 4 ft. (1.2m) power adaptor cord with NEMA 1-15 plug
240VAC models include:
3 in. (76mm) lead wire with no plug
Certifications
CSA and CE marked
Compliance
RoHS compliant
Specifications
CSA, CE, RoHS | |
10 A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.