Learn More
Bromoacetaldehyde diethyl acetal, 97%
Description
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ?-heterosubstituted acetaldehyde acetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bromoacetaldehyde diethyl acetal, is used as a synthetic building block. It is mainly used for synthesis of antibiotics such as erythromycin and cephalosporins and to other drugs. It also can be used as pharmaceutical intermediates. Besides, this chemical can be used to pruduce drugs such as methylthio imidazole, chlorpheniramine and others for thyroid. It also acts as a Useful intermediate for the preparation of other ɑ-heterosubstituted acetaldehyde acetals.
Solubility
It is immiscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Protect from frost, heat and sunlight. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Acids. Moisture Sensitive.
Specifications
Specifications
| Density | 1.27 |
| Boiling Point | 66°C to 67°C (18 mmHg) |
| Flash Point | 61°C (142°F) |
| Refractive Index | 1.439 |
| Linear Formula | BrCH2CH(OCH2CH3)2 |
| Quantity | 2500 g |
| UN Number | UN2810 |
| Beilstein | 635754 |
| Sensitivity | Moisture sensitive |
| Solubility Information | It is immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H301-H315-H319-H330 |
| Hazard Statement | GHS H Statement H301-H330-H227-H315-H319-H335 Toxic if swallowed. Fatal if inhaled. Combustible liquid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 217-989-1 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.