Learn More
G-Biosciences Bsu DNA Polymerase, Large Fragment
Bsu DNA Polymerase, Large fragment is used for second strand cDNA synthesis, strand displacement DNA synthesis, random primer labeling and dA-tailing.
$52.00 - $354.50
Description
- Bsu DNA Polymerase Large fragment is from Bacillus subtilis and is modified to retain its 5' - 3' DNA polymerase activity with its 5' - 3' exonuclease domain removed.
- It also lacks the 3' - 5' exonuclease activity.
Specifications
Standard | |
DNA Polymerase |