Learn More
Bullard™ Free-Air EDP30 Electric Driven Pumps
Supports climate control devices
Specifications
Applications | Support Climate Control Devices |
---|---|
Product Type | Electric Driven Air Pump |
Catalog Number | Mfr. No. | Horsepower | ElectricalRequirements | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Horsepower | ElectricalRequirements | Price | Quantity & Availability | ||||
17-604-203
![]() |
Bullard™
EDP30A208 |
N/A | N/A | N/A | |||||
17-604-206
![]() |
Bullard™
EDP30B230 |
N/A | N/A | N/A | |||||
17-604-205
![]() |
Bullard™
EDP30B208 |
N/A | N/A | N/A | |||||
17-604-204
![]() |
Bullard™
EDP30A230 |
N/A | N/A | N/A | |||||
17-604-207
![]() |
Bullard™
EDP30B460 |
N/A | N/A | N/A | |||||
17-604-208
![]() |
Bullard™
EDP30C230 |
N/A | N/A | N/A | |||||
17-604-209
![]() |
Bullard™
EDP30C460 |
N/A | N/A | N/A | |||||
Description
Shipped complete with inlet and outlet filter cartridges, pressure gauges, pressure relief valves, and one air supply hose quick-disconnect pod with six 0.5 in. Industrial Interchange (Hansen compatible) quick-disconnect couplers.
- Supplies air for four hood-style, six half or full face mask airline respirators, or two hood-style respirators equipped with Frigitron climate control devices
- Respirators must be approved to operate on 30 psig or less
- Easy operation and low maintenance
- Completely mobile and portable
- No expensive installation costs
Specifications
Support Climate Control Devices | |
Electric Driven Air Pump |