Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf78 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16007820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16007820 20 μL
NBP160078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16007820 Supplier Novus Biologicals Supplier No. NBP16007820UL

Rabbit Polyclonal Antibody

C17orf78 Polyclonal specifically detects C17orf78 in Human samples. It is validated for Western Blot.

Specifications

Antigen C17orf78
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N4C9
Gene Alias chromosome 17 open reading frame 78, FLJ39647, hypothetical protein LOC284099, MGC34759
Gene Symbols C17ORF78
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C17ORF78 The peptide sequence was selected from the middle region of C17ORF78. Peptide sequence LGARKLCQCQWLWRWQKKGGQPPGTAESKPDSQPQKVGQDAANSSNPKKA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284099
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.