Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C2orf55 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | C2orf55 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17046820
![]() |
Novus Biologicals
NBP17046820UL |
20 μL |
Each for $158.00
|
|
|||||
NBP170468
![]() |
Novus Biologicals
NBP170468 |
100 μL |
Each for $487.50
|
|
|||||
Description
C2orf55 Polyclonal specifically detects C2orf55 in Human samples. It is validated for Western Blot.Specifications
C2orf55 | |
Polyclonal | |
Rabbit | |
chromosome 2 open reading frame 55, hypothetical protein LOC343990, KIAA1211L, KIAA1211-like | |
KIAA1211L | |
IgG | |
102 kDa |
Western Blot | |
Unconjugated | |
RUO | |
343990 | |
Synthetic peptides corresponding to C2ORF55 The peptide sequence was selected from the middle region of C2ORF55. Peptide sequence ITVTRQKRRGTLDQPPNQEDKPGARTLKSEPGKQAKVPERGQEPVKQADF. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title