Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-16, Mouse anti-Human, Clone: CDH16/2125, Novus Biologicals™
SDP

Catalog No. p-7182555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010462 20 μg
NB010461 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010462 Supplier Novus Biologicals Supplier No. NBP27972220UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-16 Monoclonal specifically detects Cadherin-16 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Cadherin-16
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CDH16/2125
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. O75309
Gene Alias cadherin 16, KSP-cadherin, cadherin-16, Kidney-specific cadherin, KSP-cadherin
Gene Symbols CDH16
Host Species Mouse
Immunogen Recombinant fragment (around aa 371-507) of human Cadherin-16 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1014
Test Specificity This monoclonal antibody recognizes a protein of 130kDa, identified as Cadherin-16; Cadherin-16 has been suggested to distinguish Chromophobe Renal-Cell Carcinoma from Oncocytoma.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.