Description
- Standard in Moh's scale of hardness (3)
- Example of three-plane rhombic cleavage
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
| Product Type | Mineral, Calcite |
| For Use With (Application) | Study on mineral hardness, cleavage, light refraction, and effervescence |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 1 Ea. |
| Specimen Size | 2 to 3 cm |