Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcium Green™-1, AM, cell permeant

Catalog No. c3012
Encompass_Preferred
Click to view available options
Quantity:
10 x 50μg
500μg

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

  • They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
  • Cells may be loaded with the AM ester forms of these calcium indicators by adding dissolved indicator directly to dishes containing cultured cells
  • Fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
  • Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem

  • Label (Ex/Em): Calcium Green; -1 (506/531nm)
  • Fluorescence intensity increase upon binding Ca2+ : approximately 14 fold

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Fluorescent Dye-Based
Product Line Calcium Green
Quantity 10 x 50μg
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.