Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium-sensing R/CaSR Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200075584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435932 0.025 mg
NBP241271 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435932 Supplier Novus Biologicals Supplier No. NBP2412710.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcium-sensing R/CaSR Polyclonal specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Calcium-sensing R/CaSR
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence
Formulation PBS with 0.02% Sodium Azide
Gene Alias calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Gene Symbols CASR
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the amino terminus of human CASR. The immunogen is located within the first 50 amino acids of CASR. Amino Acid Squence: TSAYGPDQRAQKKGD
Molecular Weight of Antigen 115, 125 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 846
Test Specificity At least two isoforms of CASR are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.