Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMP/Cathelicidin Antibody (OSX12), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N146781APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N146781APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N146781APC7 Supplier Novus Biologicals Supplier No. NBP146781APCCY7

Mouse Monoclonal Antibody

CAMP/Cathelicidin Monoclonal specifically detects CAMP/Cathelicidin in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CAMP/Cathelicidin
Applications Flow Cytometry
Classification Monoclonal
Clone OSX12
Conjugate APC-Cyanine7
Dilution Flow Cytometry 5-10 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37
Gene Symbols CAMP
Host Species Mouse
Immunogen A synthetic peptide from human CAMP/Cathelicidin conjugated to an immunogenic carrier protein has been used as the antigen.
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 820.0
Test Specificity Specific for Cathelicidin antimicrobial peptide.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.