Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-Sala™ Delta™ Vest Style Harnesses
No-Tangle™ triangular design creates frame that allows the harness to fall into place when donned.
$287.67 - $338.27
Catalog Number | Mfr. No. | Size | D-Ring Location | Buckle Style (Leg) | Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | D-Ring Location | Buckle Style (Leg) | Style | Price | Quantity & Availability | ||
19-066-803
|
3M™
7012815465 |
Universal | 1 back, 2 hip | Pass-Through | Vest Style |
Each for $315.71
|
|
||
19-066-800
|
3M™
7012691269 |
Universal | 1 back, 2 hip | Tongue Buckle | Vest Style |
Each for $338.27
|
|
||
19-066-884
|
3M™
7012691234 |
X-Large | Back, Side | Tongue Buckle | Vest Style |
Each for $338.27
|
|
||
19-066-804
|
3M™
7012815462 |
Universal | 1 back | Pass through | Vest Style |
Each for $287.67
|
|
||
18-706
|
3M™
1103512 |
Universal | 1 back, 2 hip | Pass-Through | Construction Style, w/ loops for belt |
Each for $291.30
|
|
||
Description
- Spring-loaded stand-up dorsal D-ring design ensures fast, easy and safe connections
- Rip-stitch indicators allow to inspect the harness at glance, for prior damaging impact loads
- Harness falls into place, allowing quick and easy donning
- Parachute buckles on lower shoulder straps for quick and easy adjustment
- Side D-rings for positioning
- Adjustable non-slip chest strap with easy to use pass-thru buckle
- Tongue buckle leg straps for ease of use and will not loose
- Lanyard keepers holds unused snap hooks and help reduce trip hazard
- Polyester web construction to add durability and forged alloy steel hardware
- Revolver vertical torso adjuster provides simple and fast adjustment eliminating loose ends and locks webbing into place preventing slippage
- Velcro cover protects harness labels and holds I-Safe™ ID tag
- Capacity: 420 lb. (190.5kg)
Compliance
Meets ANSI™ Z359.11 requirements.
Specifications
Pass-Through |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title