Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbopack X™ Thermal Desorption Tubes

Catalog No. 111005354 Shop All MilliporeSigma Supelco Products
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps

Suitable for use with Air Monitoring

  • For analyte group C3-C5
TRUSTED_SUSTAINABILITY

Specifications

Matrix Carbopack™ x Graphitized Carbon Black (GCB)
Material Glass
Methods EPA TO-17
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Description Pre-conditioned/Sealed with Brass Endcaps
Packaging Quantity 10/Pk.
Product Line Carbopack
For Use With (Application) Air Monitoring
Diameter (English) Outer 1/4 in.
Overall Length (English) 3.5 in.
Type Thermal Desorption Tube
Show More Show Less

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.