Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbopack X™ Thermal Desorption Tubes

Catalog No. p-7202876
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps

Suitable for use with Air Monitoring

  • For analyte group C3-C5
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Material Glass
Product Line Carbopack
Matrix Carbopack™ x Graphitized Carbon Black (GCB)
Diameter (English) Outer 1/4 in.
Packaging Quantity 10/Pk.
Overall Length (English) 3.5 in.
For Use With (Application) Air Monitoring
Methods EPA TO-17
Description Pre-conditioned
Type Thermal Desorption Tube
Show More Show Less

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.