Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbosieve™ SIII Thermal Desorption Tubes

Catalog No. 111006687 Shop All MilliporeSigma Supelco Products
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps

Suitable for use with Air Monitoring

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Air Monitoring
Overall Length (English) 3.5 in.
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Packaging Quantity 10/Pk.
Material Stainless Steel
Description Pre-conditioned/Sealed with Brass Endcaps
Product Line Carbosieve
Diameter (English) Outer 1/4 in.
Matrix Carbosieve™ S-III Carbon Molecular Sieve
Methods EPA TO-2
Type Desorption Tube
Show More Show Less

Carbosieve is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.