Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carnosine Dipeptidase 1/CNDP1 Antibody (001), PerCP, Novus Biologicals™
SDP

Catalog No. NB308189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308189 Supplier Novus Biologicals Supplier No. NBP289263PCP
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carnosine Dipeptidase 1/CNDP1 Monoclonal antibody specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Carnosine Dipeptidase 1/CNDP1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 001
Conjugate PerCP
Dilution ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carnosine Dipeptidase 1/CNDP1 (Accession#: NP_116038.4; Ser27-His507).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84735
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.