Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASKIN2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB341666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341666 Supplier Novus Biologicals Supplier No. NBP281971MFV500
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASKIN2 Polyclonal antibody specifically detects CASKIN2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CASKIN2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ANKS5B, CASK interacting protein 2, caskin-2, FLJ21609, KIAA1139
Host Species Rabbit
Immunogen CASKIN2 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human CASKIN2. The immunogen is located within amino acids 380 - 430 of CASKIN2. Amino Acid Squence: RVGLSPDSPAGDRNSVGSE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57513
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.