Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-2 Antibody (SR1824), Novus Biologicals™
SDP

Catalog No. NB260521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260521 100 μg
NB258741 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260521 Supplier Novus Biologicals Supplier No. NBP321672100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Caspase-2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1824
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57
Host Species Rabbit
Immunogen A synthesized peptide derived from human Caspase-2 (Uniprot #: P42575)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 835
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.