Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-9 Antibody, Novus Biologicals™
SDP

Catalog No. p-5053843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056118T 0.025 mL
NB10056118 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056118T Supplier Novus Biologicals Supplier No. NB10056118SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 19 publications

Caspase-9 Polyclonal specifically detects Caspase-9 in Human, Mouse, Rat, Canine, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Specifications

Antigen Caspase-9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 24184144), Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22223358), Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen Assay Dependent, Flow (Intracellular)
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. P55211
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbols CASP9
Host Species Rabbit
Immunogen Human Caspase 9 recombinant protein catalytic subunits (NP_001220).
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 842
Test Specificity This antibody detects both the pro- and active-Caspase 9 forms. In western blots, the proform of caspase-9 is detected at ∼50 kDa, the large subunit at ∼35 kDa, and the small subunit at ∼15 kDa. Intermediate caspase-9 cleavage forms may also be seen at ∼21 kDa.
Target Species Human, Mouse, Rat, Canine, Gerbil
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.