Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CATSPERE Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $548.00
Specifications
Antigen | C1orf101 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23784920
![]() |
Novus Biologicals
NBP23784920UL |
20 μL |
Each for $158.00
|
|
|||||
NBP237849
![]() |
Novus Biologicals
NBP237849 |
100 μL |
Each for $548.00
|
|
|||||
Description
CATSPERE Polyclonal specifically detects CATSPERE in Human samples. It is validated for Western Blot.Specifications
C1orf101 | |
Polyclonal | |
Rabbit | |
Human | |
257044 | |
IgG | |
91 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q5SY80.2 | |
C1orf101 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title