Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL7/MCP-3/MARC Antibody (36320), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348704 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348704 Supplier Novus Biologicals Supplier No. FAB282MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL7/MCP-3/MARC Monoclonal antibody specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF

Specifications

Antigen CCL7/MCP-3/MARC
Applications Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 36320
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3)
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL7/MCP-3/MARC, Gln34-109, Accession # Q7Z7Q8
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6354
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.