Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCNB3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CCNB3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1802720
![]() |
Novus Biologicals
NBP18012720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180127
![]() |
Novus Biologicals
NBP180127 |
100 μL |
Each for $487.50
|
|
|||||
Description
CCNB3 Polyclonal specifically detects CCNB3 in Human samples. It is validated for Western Blot.Specifications
CCNB3 | |
Polyclonal | |
Purified | |
RUO | |
NP_391990 | |
85417 | |
Synthetic peptide directed towards the N terminal of human CCNB3. Peptide sequence: MLLPLPPQSSKPVPKKSQSSKIVPSHHDPSEKTGENCQTKISPSSLQESP | |
Primary | |
291 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
CYCB3, cyclin B3, G2/mitotic-specific cyclin-B3 | |
CCNB3 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title