Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCT8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | CCT8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15660820
![]() |
Novus Biologicals
NBP15660820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156608
![]() |
Novus Biologicals
NBP156608 |
100 μL |
Each for $487.50
|
|
|||||
Description
CCT8 Polyclonal specifically detects CCT8 in Human, Mouse samples. It is validated for Western Blot.Specifications
CCT8 | |
Polyclonal | |
Rabbit | |
C21orf112, CCTQ, CCT-theta, chaperonin containing TCP1, subunit 8 (theta), chromosome 21 open reading frame 112, D21S246, KIAA0002, PRED71, T-complex protein 1 subunit theta, T-complex protein 1, theta subunit, 10Renal carcinoma antigen NY-REN-15, TCP-1-theta | |
CCT8 | |
IgG | |
This product is specific to Subunit or Isoform: theta. |
Western Blot | |
Unconjugated | |
RUO | |
10694 | |
Synthetic peptides corresponding to CCT8(chaperonin containing TCP1, subunit 8 (theta)) The peptide sequence was selected from the C terminal of CCT8. Peptide sequence DMLEAGILDTYLGKYWAIKLATNAAVTVLRVDQIIMAKPAGGPKPPSGKK. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title