Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD120b (TNF Receptor II) Monoclonal Antibody (TR75-54), eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

$123.00 - $288.00

Specifications

Antigen CD120b (TNF Receptor II)
Clone TR75-54
Concentration 0.5 mg/mL
Applications ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-128-36
Encompass_Preferred
View Documents
Invitrogen™
14120381
50 μg
Each for $123.00

Save 25%

Reg : $164.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
50-128-37
Encompass_Preferred
View Documents
Invitrogen™
14120385
500 μg
Each for $288.00

Save 25%

Reg : $384.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
Description

Description

Description: The TR75-54 monoclonal antibody reacts with mouse Tumor Necrosis Factor Receptor II (TNFRII, TNFR-p80, TNFRSF1B, CD120b). TNFRII is expressed in a variety of cell types and strongly expressed on stimulated T and B lymphocytes. TNFRII is the main TNF receptor found on circulating T cells and is the major mediator of autoregulatory apoptosis in CD8+ cells. The soluble TNF receptors are truncated forms of cell surface receptors with neutralizing activity on both TNF-alpha and TNF-beta. It has been suggested that the proinflammatory and immunosuppressive properties of TNF segregate at the level of its receptors and that the pool of TNF bound to soluble receptors could represent a reservoir to control the TNF release. Applications Reported: This TR75-54 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and ELISA. Applications Tested: The TR75-54 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse TNFRII in combination with the biotin TR75-32 (cat. 13-1204) antibody for detection and recombit mouse TNFRII as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE.

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Specifications

Specifications

CD120b (TNF Receptor II)
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Tnfrsf1b
Primary
4°C
Tnfrsf1b
TR75-54
ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Unconjugated
Armenian Hamster
Mouse
P25119
21938
IgG
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.